missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-hydroxyphenylacetic acid, 98%

Catalog No. p-7081919
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1211403 1 g
AAA1211406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1211403 Supplier Thermo Scientific Chemicals Supplier No. A1211403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 458-09-3 | C8H7FO3 | 170.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 458-09-3
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00004348
InChI Key YRFBZAHYMOSSGX-UHFFFAOYSA-N
Synonym 3-fluoro-4-hydroxyphenylacetic acid, 2-3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxybenzeneacetic acid, benzeneacetic acid, 3-fluoro-4-hydroxy, 2-3-fluoro-4-hydroxy-phenyl acetic acid, 4-carboxymethyl-2-fluorophenol, 3-fluoro-4-hydroxy-phenyl-acetic acid, acmc-1ahjh, opera_id_1839
PubChem CID 68014
IUPAC Name 2-(3-fluoro-4-hydroxyphenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)F)O

Specifications

Melting Point 130°C to 133°C
Quantity 1 g
Formula Weight 170.14
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-hydroxyphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-275-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only