missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-hydroxybenzaldehyde 98.0+%, TCI America™

Catalog No. F07251G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07251G 1 g
F07255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07251G Supplier TCI America Supplier No. F07251G

Chemical Identifiers

CAS 405-05-0
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD00016626
InChI Key QSBHJTCAPWOIIE-UHFFFAOYSA-N
Synonym 3-fluoro-4-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-4-hydroxy, 4-hydroxy-3-fluorobenzaldehyde, pubchem1443, acmc-1acq8, ksc497m8t, 3-fluoro-4 hydroxybenzaldehyde, 4-hydroxy-3-fluoro-benzaldehyde, #, 3-fluoranyl-4-oxidanyl-benzaldehyde
PubChem CID 587250
IUPAC Name 3-fluoro-4-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1C=O)F)O

Specifications

Melting Point 125°C
Quantity 1 g
Formula Weight 140.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-hydroxybenzaldehyde