accessibility menu, dialog, popup

UserName

CAS RN 405-05-0

IUPAC Name: 3-fluoro-4-hydroxybenzaldehyde
Synonyms: 3-fluoro-4-hydroxybenzaldehyde
Molecular Weight (g/mol): 140.11
Molecular Formula: C7H5FO2
InChi Key: QSBHJTCAPWOIIE-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(C=O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)