missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-hydroxy-5-methoxybenzaldehyde, 96%

Catalog No. AAH66477MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66477MD 250 mg
AAH6647703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66477MD Supplier Thermo Scientific Chemicals Supplier No. H66477MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79418-78-3 | C8H7FO3 | 170.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79418-78-3
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD02683560
InChI Key OOGOFUKAJDPHDJ-UHFFFAOYSA-N
Synonym 5-fluorovanillin, benzaldehyde, 3-fluoro-4-hydroxy-5-methoxy, 3-fluoro-4-hydroxy-5-methoxy-benzaldehyde, pubchem10464, acmc-209xkq, 3-fluoro-4-hydroxy-5-methoxy benzaldehyde, 3-fluoro-5-methoxy-4-hydroxybenzaldehyde, #, 3-fluoranyl-5-methoxy-4-oxidanyl-benzaldehyde, 3-fluoro-4-hydroxy-5-methyloxy benzaldehyde
PubChem CID 598452
IUPAC Name 3-fluoro-4-hydroxy-5-methoxybenzaldehyde
SMILES COC1=C(C(=CC(=C1)C=O)F)O

Specifications

Color Brown
Boiling Point 114°C to 118°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 170.14
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 3-Fluoro-4-hydroxy-5-methoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only