missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-formylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F10511G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10511G Supplier TCI America Supplier No. F10511G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Fluoro-4-formylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 248270-25-9
Melting Point 240°C
Color White-Yellow
Molecular Formula C7H6BFO3
Quantity 1 g
MDL Number MFCD02093051
Synonym 3-fluoro-4-formylbenzeneboronic acid, 3-fluoro-4-formylphenyl boronic acid, 3-fluoro-4-formylphenboronic acid, boronic acid, 3-fluoro-4-formylphenyl, pubchem1847, acmc-1cga0, ksc494k4h, 4-borono-3-fluorobenzaldehyde, 3-fluoro-4-formyl-phenyl boronic acid, 3-fluoranyl-4-methanoyl-phenyl boronic acid
InChI Key NZNRMUVHUVCIBR-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)C=O)F)(O)O
IUPAC Name (3-fluoro-4-formylphenyl)boronic acid
Molecular Weight (g/mol) 167.93
PubChem CID 2778654
Formula Weight 167.93
Physical Form Crystalline Powder
Show More Show Less