missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%

CAS: 1408088-34-5 | C17H26BFN2O2 | 320.215 g/mol

$208.30 - $587.85

Chemical Identifiers

CAS 1408088-34-5
Molecular Formula C17H26BFN2O2
Molecular Weight (g/mol) 320.215
MDL Number MFCD22988988
InChI Key RFECQDXNBWGCRR-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-4-methylpiperazine, 3-fluoro-4-4-methyl-1-piperazinyl benzeneboronic acid pinacol ester
PubChem CID 70974710
IUPAC Name 1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCN(CC3)C)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59640MD
View Documents
Thermo Scientific Chemicals
H59640MD
250 mg
Each for $208.30
Only null left
 
AAH5964003
View Documents
Thermo Scientific Chemicals
H5964003
1 g
Each for $587.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

1408088-34-5
320.215
RFECQDXNBWGCRR-UHFFFAOYSA-N
70974710
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCN(CC3)C)F
C17H26BFN2O2
MFCD22988988
1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-4-methylpiperazine, 3-fluoro-4-4-methyl-1-piperazinyl benzeneboronic acid pinacol ester
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine

Specifications

1408088-34-5
MFCD22988988
1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-4-methylpiperazine, 3-fluoro-4-4-methyl-1-piperazinyl benzeneboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCN(CC3)C)F
320.215
320.21
3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester
C17H26BFN2O2
250 mg
RFECQDXNBWGCRR-UHFFFAOYSA-N
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine
70974710
95%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only