CAS RN 1408088-34-5
IUPAC Name:
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine
Synonyms:
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine
Molecular Weight (g/mol):
320.22
Molecular Formula:
C17H26BFN2O2
InChi Key:
RFECQDXNBWGCRR-UHFFFAOYSA-N
SMILES:
CN1CCN(CC1)C1=C(F)C=C(C=C1)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
Promotions Available
3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%
3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%
CAS: 1408088-34-5 Molecular Formula: C17H26BFN2O2 Molecular Weight (g/mol): 320.215 MDL Number: MFCD22988988 InChI Key: RFECQDXNBWGCRR-UHFFFAOYSA-N Synonym: 1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-4-methylpiperazine,3-fluoro-4-4-methyl-1-piperazinyl benzeneboronic acid pinacol ester PubChem CID: 70974710 IUPAC Name: 1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCN(CC3)C)F