missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%

Catalog No. AAH59640MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59640MD 250 mg
AAH5964003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59640MD Supplier Thermo Scientific Chemicals Supplier No. H59640MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1408088-34-5 | C17H26BFN2O2 | 320.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1408088-34-5
Molecular Formula C17H26BFN2O2
Molecular Weight (g/mol) 320.215
MDL Number MFCD22988988
InChI Key RFECQDXNBWGCRR-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-4-methylpiperazine, 3-fluoro-4-4-methyl-1-piperazinyl benzeneboronic acid pinacol ester
PubChem CID 70974710
IUPAC Name 1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCN(CC3)C)F

Specifications

Quantity 250 mg
Formula Weight 320.21
Percent Purity 95%
Chemical Name or Material 3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only