missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methylpyridine, 98%

Catalog No. p-7055978
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6470503 1 g
AAH6470506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6470503 Supplier Thermo Scientific Chemicals Supplier No. H6470503

CAS: 15931-15-4 | C6H6FN | 111.119 g/mol

3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 15931-15-4
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD08705710
InChI Key IRIHSZDBTACXCT-UHFFFAOYSA-N
Synonym 3-fluoro-2-picoline, pyridine, 3-fluoro-2-methyl, 2-methyl-3-fluoropyridine, 3-fluoro-2-methyl pyridine, 3-fluoro-2-methyl-pyridine, fluoropicoline, pubchem14381, acmc-209xci, ksc530a3b, pyridine,3-fluoro-2-methyl
PubChem CID 21613172
IUPAC Name 3-fluoro-2-methylpyridine
SMILES CC1=C(C=CC=N1)F

Specifications

Color Colorless to Yellow
Quantity 1 g
Beilstein 6497832
Solubility Information Slightly soluble in water.
Formula Weight 111.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Fluoro-2-methylpyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only