missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methylaniline 98.0+%, TCI America™

Catalog No. F05265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05265G 5 g
F052625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05265G Supplier TCI America Supplier No. F05265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 443-86-7
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007760
InChI Key SLDLVGFPFFLYBM-UHFFFAOYSA-N
Synonym 3-fluoro-o-toluidine, 2-fluoro-6-aminotoluene, 2-amino-6-fluorotoluene, 6-amino-2-fluorotoluene, 3-fluoro-2-methyl-aniline, benzenamine, 3-fluoro-2-methyl, 3-fluoro-2-methylphenylamine, 2-methyl-3-fluoroaniline, hydrochloride, 3-fluor-2-methylanilin
PubChem CID 285553
IUPAC Name 3-fluoro-2-methylaniline
SMILES CC1=C(N)C=CC=C1F

Specifications

Boiling Point 205°C
Quantity 5 g
Formula Weight 125.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Fluoro-2-methylaniline