missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methoxy-6-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog No. p-7055961
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6468306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6468306 Supplier Thermo Scientific Chemicals Supplier No. H6468306
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 375368-80-2
Molecular Formula C7H8FNO
Molecular Weight (g/mol) 141.15
MDL Number MFCD03095083
InChI Key DAQMWRIPXIPMAA-UHFFFAOYSA-N
Synonym 3-fluoro-2-methoxy-6-picoline, 2-methoxy-3-fluoro-6-methylpyridine, 3-fluoro-2-methoxy-6-methyl-pyridine, pubchem6585, ksc914i9l, abbypharma ap-15-5217, pyridine, 3-fluoro-2-methoxy-6-methyl, pyridine, 3-fluoro-2-methoxy-6-methyl-9ci
PubChem CID 24820319
IUPAC Name 3-fluoro-2-methoxy-6-methylpyridine
SMILES COC1=C(F)C=CC(C)=N1

Specifications

Color Colorless to Yellow
Quantity 5 g
Formula Weight 141.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Fluoro-2-methoxy-6-methylpyridine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only