missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-hydroxypyridine, 97%

CAS: 1547-29-1 | C5H4FNO | 113.091 g/mol

Supplier:  Thermo Scientific Chemicals H3170506

 View more versions of this product

Catalog No. AAH3170506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Fluoro-2-hydroxypyridine
157°C to 161°C
C5H4FNO
5 g
CISUDJRBCKKAGX-UHFFFAOYSA-N
3-fluoro-1H-pyridin-2-one
2783363
97%
1547-29-1
Odorless
MFCD03095042
3-fluoro-2-hydroxypyridine, 3-fluoropyridin-2-ol, 3-fluoro-2-pyridinol, 2 1h-pyridinone, 3-fluoro, 3-fluoropyridin-2 1h-one, 3-fluoro-pyridin-2-ol, 3-fluoro-2-pyridone, 3-fluoro-2-hydroxypyridine, 3-fluoro-2-pyridone, fluoropyridone, pubchem1200
C1=CNC(=O)C(=C1)F
113.091
113.09

Chemical Identifiers

1547-29-1
113.091
CISUDJRBCKKAGX-UHFFFAOYSA-N
2783363
C1=CNC(=O)C(=C1)F
C5H4FNO
MFCD03095042
3-fluoro-2-hydroxypyridine, 3-fluoropyridin-2-ol, 3-fluoro-2-pyridinol, 2 1h-pyridinone, 3-fluoro, 3-fluoropyridin-2 1h-one, 3-fluoro-pyridin-2-ol, 3-fluoro-2-pyridone, 3-fluoro-2-hydroxypyridine, 3-fluoro-2-pyridone, fluoropyridone, pubchem1200
3-fluoro-1H-pyridin-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only