CAS RN 1547-29-1
IUPAC Name:
3-fluoro-1,2-dihydropyridin-2-one
Synonyms:
3-fluoro-1H-pyridin-2-one
Molecular Weight (g/mol):
113.09
Molecular Formula:
C5H4FNO
InChi Key:
CISUDJRBCKKAGX-UHFFFAOYSA-N
SMILES:
FC1=CC=CNC1=O
1
–
1
of
1
results
1
Promotions Available
3-Fluoro-2-hydroxypyridine, 97%
3-Fluoro-2-hydroxypyridine, 97%
CAS: 1547-29-1 Molecular Formula: C5H4FNO Molecular Weight (g/mol): 113.091 MDL Number: MFCD03095042 InChI Key: CISUDJRBCKKAGX-UHFFFAOYSA-N Synonym: 3-fluoro-2-hydroxypyridine,3-fluoropyridin-2-ol,3-fluoro-2-pyridinol,2 1h-pyridinone, 3-fluoro,3-fluoropyridin-2 1h-one,3-fluoro-pyridin-2-ol,3-fluoro-2-pyridone,3-fluoro-2-hydroxypyridine, 3-fluoro-2-pyridone,fluoropyridone,pubchem1200 PubChem CID: 2783363 IUPAC Name: 3-fluoro-1H-pyridin-2-one SMILES: C1=CNC(=O)C(=C1)F