missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-hydroxypyridine, 97%

Catalog No. AAH31705MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31705MD 250 mg
AAH3170503 1 g
AAH3170506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH31705MD Supplier Thermo Scientific Chemicals Supplier No. H31705MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1547-29-1 | C5H4FNO | 113.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1547-29-1
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.091
MDL Number MFCD03095042
InChI Key CISUDJRBCKKAGX-UHFFFAOYSA-N
Synonym 3-fluoro-2-hydroxypyridine, 3-fluoropyridin-2-ol, 3-fluoro-2-pyridinol, 2 1h-pyridinone, 3-fluoro, 3-fluoropyridin-2 1h-one, 3-fluoro-pyridin-2-ol, 3-fluoro-2-pyridone, 3-fluoro-2-hydroxypyridine, 3-fluoro-2-pyridone, fluoropyridone, pubchem1200
PubChem CID 2783363
IUPAC Name 3-fluoro-1H-pyridin-2-one
SMILES C1=CNC(=O)C(=C1)F

Specifications

Melting Point 157°C to 161°C
Odor Odorless
Quantity 250 mg
Formula Weight 113.09
Percent Purity 97%
Chemical Name or Material 3-Fluoro-2-hydroxypyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only