missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-2-formylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$87.60 - $87.60
Specifications
| CAS | 871126-15-7 |
|---|---|
| Melting Point | 129°C |
| Color | White |
| Molecular Formula | C7H6BFO3 |
| MDL Number | MFCD10697421 |
Specifications
| 871126-15-7 | |
| White | |
| MFCD10697421 | |
| GVHWLCYABLWGIR-UHFFFAOYSA-N | |
| (3-fluoro-2-formylphenyl)boronic acid | |
| 53412038 | |
| Crystalline Powder |
| 129°C | |
| C7H6BFO3 | |
| 3261 | |
| B(C1=C(C(=CC=C1)F)C=O)(O)O | |
| 167.93 | |
| 167.93 |