missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-formylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F10891G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10891G Supplier TCI America Supplier No. F10891G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Fluoro-2-formylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 871126-15-7
Melting Point 129°C
Color White
Molecular Formula C7H6BFO3
Quantity 1 g
MDL Number MFCD10697421
UN Number 3261
Synonym 3-fluoro-2-formylphenyl boronic acid, 3-fluoro-2-formylbenzeneboronic acid, boronic acid, 3-fluoro-2-formylphenyl, acmc-209qf3, 2-formyl-3-fluorophenylboranic acid, 3-fluoro-2-formyl-phenyl boronic acid, 3-fluoro-2-formylphenylboronic acid, boronic acid, b-3-fluoro-2-formylphenyl, contains varying amounts of anhydride
InChI Key GVHWLCYABLWGIR-UHFFFAOYSA-N
SMILES B(C1=C(C(=CC=C1)F)C=O)(O)O
IUPAC Name (3-fluoro-2-formylphenyl)boronic acid
Molecular Weight (g/mol) 167.93
PubChem CID 53412038
Formula Weight 167.93
Physical Form Crystalline Powder
Show More Show Less