missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-formylbenzeneboronic acid, 95%

Catalog No. p-7056146
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6490903 1 g
AAH6490906 5 g
AAH6490914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6490903 Supplier Thermo Scientific Chemicals Supplier No. H6490903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 871126-15-7 | C7H6BFO3 | 167.93 g/mol

Undergoes unprecedented tautomeric rearrangement in solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Undergoes unprecedented tautomeric rearrangement in solution.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.

Chemical Identifiers

CAS 871126-15-7
Molecular Formula C7H6BFO3
Molecular Weight (g/mol) 167.93
MDL Number MFCD10697421
InChI Key GVHWLCYABLWGIR-UHFFFAOYSA-N
Synonym 3-fluoro-2-formylphenyl boronic acid, 3-fluoro-2-formylbenzeneboronic acid, boronic acid, 3-fluoro-2-formylphenyl, acmc-209qf3, 2-formyl-3-fluorophenylboranic acid, 3-fluoro-2-formyl-phenyl boronic acid, 3-fluoro-2-formylphenylboronic acid, boronic acid, b-3-fluoro-2-formylphenyl, contains varying amounts of anhydride
PubChem CID 53412038
IUPAC Name (3-fluoro-2-formylphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)F)C=O)(O)O

Specifications

Melting Point 123°C to 127°C
Quantity 1 g
UN Number UN3261
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 167.93
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Fluoro-2-formylbenzeneboronic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only