missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-1,2-dihydroxybenzene, 98+%, Thermo Scientific Chemicals
$83.66 - $83.66
Chemical Identifiers
| CAS | 363-52-0 |
|---|---|
| Molecular Formula | C6H5FO2 |
| Molecular Weight (g/mol) | 128.10 |
| MDL Number | MFCD00042582 |
| InChI Key | DXOSJQLIRGXWCF-UHFFFAOYSA-N |
| Synonym | 3-fluorocatechol, pyrocatechol, 3-fluoro, 1,2-benzenediol, 3-fluoro, 1,2-dihydroxy-3-fluorobenzene, 3-fluoro-1,2-dihydroxybenzene, 3-fluoro-1,2-benzenediol, 1-fluoro-2,3-dihydroxybenzene, 3fa, pubchem4142 |
| PubChem CID | 67764 |
| ChEBI | CHEBI:39876 |
| IUPAC Name | 3-fluorobenzene-1,2-diol |
| SMILES | OC1=CC=CC(F)=C1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC264790010
|
Thermo Scientific Chemicals
264790010 |
1 g | Glass bottle |
Each for $83.66
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 363-52-0 | |
| 128.10 | |
| DXOSJQLIRGXWCF-UHFFFAOYSA-N | |
| 67764 | |
| 3-fluorobenzene-1,2-diol |
| C6H5FO2 | |
| MFCD00042582 | |
| 3-fluorocatechol, pyrocatechol, 3-fluoro, 1,2-benzenediol, 3-fluoro, 1,2-dihydroxy-3-fluorobenzene, 3-fluoro-1,2-dihydroxybenzene, 3-fluoro-1,2-benzenediol, 1-fluoro-2,3-dihydroxybenzene, 3fa, pubchem4142 | |
| CHEBI:39876 | |
| OC1=CC=CC(F)=C1O |
Specifications
| 363-52-0 | |
| 100.0 | |
| Beige to Brown-Gray | |
| 98+% | |
| C6H5FO2 | |
| 1 g | |
| Solubility in water: >350g/L (20°C). Other solubilities: soluble in alcohols,ketones,esters | |
| OC1=CC=CC(F)=C1O | |
| 128.10 | |
| CHEBI:39876 | |
| 98+% | |
| 3-Fluoro-1, 2-dihydroxybenzene, 98+% |
| 98.0 | |
| 71°C to 74°C | |
| Authentic | |
| Glass bottle | |
| MFCD00042582 | |
| 3-fluorocatechol, pyrocatechol, 3-fluoro, 1,2-benzenediol, 3-fluoro, 1,2-dihydroxy-3-fluorobenzene, 3-fluoro-1,2-dihydroxybenzene, 3-fluoro-1,2-benzenediol, 1-fluoro-2,3-dihydroxybenzene, 3fa, pubchem4142 | |
| DXOSJQLIRGXWCF-UHFFFAOYSA-N | |
| 3-fluorobenzene-1,2-diol | |
| 67764 | |
| 128.1 | |
| Crystalline Powder |
Safety and Handling
TSCA : TSCA
RUO – Research Use Only