missing translation for 'onlineSavingsMsg'
Learn More

3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide, 99+%, Thermo Scientific Chemicals

Catalog No. AC277810010
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC277810010 1 g Glass Bottle
AC277812500 250 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC277810010 Supplier Thermo Scientific Chemicals Supplier No. 277810010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105786-40-1
Molecular Formula C8H13N2O
Molecular Weight (g/mol) 153.20
MDL Number MFCD00143157,MFCD01456476,MFCD00143157
InChI Key SCQSHSJVMGGQKR-UCROKIRRSA-O
Synonym 3-exo-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, +-1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, +/--1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, bicyclo 2.2.1 hept-5-ene-2-carboxamide, 3-amino-, 1r,2r,3s,4s-rel-9ci
PubChem CID 42580098
IUPAC Name (1S,2S,3R,4R)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxamide
SMILES NC(=O)[C@H]1[C@@H]([NH3+])[C@@H]2C[C@H]1C=C2

Specifications

Melting Point 235.0°C to 253.0°C
CAS Min % 99.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol and methanol
Formula Weight 152.2
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material 3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide, 99+%
Show More Show Less

RUO – Research Use Only