CAS RN 105786-40-1
IUPAC Name:
(1R,2S,3R,4S)-3-carbamoylbicyclo[2.2.1]hept-5-en-2-aminium
Synonyms:
(1R,2S,3R,4S)-3-carbamoylbicyclo[2.2.1]hept-5-en-2-aminium
Molecular Weight (g/mol):
153.2
Molecular Formula:
C8H13N2O
InChi Key:
SCQSHSJVMGGQKR-UCROKIRRSA-O
SMILES:
NC(=O)[C@H]1[C@@H]([NH3+])[C@@H]2C[C@H]1C=C2
1
–
1
of
1
results
1
Promotions Available
3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide, 99+%, Thermo Scientific Chemicals
3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide, 99+%, Thermo Scientific Chemicals
CAS: 105786-40-1 Molecular Formula: C8H13N2O Molecular Weight (g/mol): 153.20 MDL Number: MFCD00143157,MFCD01456476,MFCD00143157 InChI Key: SCQSHSJVMGGQKR-UCROKIRRSA-O Synonym: 3-exo-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide,+-1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide,+/--1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide,bicyclo 2.2.1 hept-5-ene-2-carboxamide, 3-amino-, 1r,2r,3s,4s-rel-9ci PubChem CID: 42580098 IUPAC Name: (1S,2S,3R,4R)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxamide SMILES: NC(=O)[C@H]1[C@@H]([NH3+])[C@@H]2C[C@H]1C=C2