missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynyltoluene 98.0+%, TCI America™

Catalog No. E06651G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06651G 1 g
E06655G 5 g
E066525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E06651G Supplier TCI America Supplier No. E06651G

Chemical Identifiers

CAS 766-82-5
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
MDL Number MFCD01318174
InChI Key RENYIDZOAFFNHC-UHFFFAOYSA-N
PubChem CID 136600
IUPAC Name 1-ethynyl-3-methylbenzene
SMILES CC1=CC=CC(=C1)C#C

Specifications

Color Yellow
Boiling Point 60°C
Quantity 1 g
UN Number 3295
Synonym 3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethynyltoluene