missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethynylthiophene 97.0+%, TCI America™
$121.28 - $348.46
Chemical Identifiers
| CAS | 67237-53-0 |
|---|---|
| Molecular Formula | C6H4S |
| Molecular Weight (g/mol) | 108.158 |
| MDL Number | MFCD04039973 |
| InChI Key | MJHLPKWONJUCFK-UHFFFAOYSA-N |
| Synonym | 3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b |
| PubChem CID | 3548422 |
| IUPAC Name | 3-ethynylthiophene |
| SMILES | C#CC1=CSC=C1 |
Chemical Identifiers
| 67237-53-0 | |
| 108.158 | |
| MJHLPKWONJUCFK-UHFFFAOYSA-N | |
| 3548422 | |
| C#CC1=CSC=C1 |
| C6H4S | |
| MFCD04039973 | |
| 3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b | |
| 3-ethynylthiophene |
Specifications
| 67237-53-0 | |
| 50°C | |
| MFCD04039973 | |
| 1993 | |
| MJHLPKWONJUCFK-UHFFFAOYSA-N | |
| 3-ethynylthiophene | |
| 3548422 | |
| ≥97.0% (GC) | |
| 3-Ethynylthiophene |
| Yellow | |
| C6H4S | |
| 1 g | |
| 3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b | |
| C#CC1=CSC=C1 | |
| 108.158 | |
| 108.16 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator