accessibility menu, dialog, popup

UserName

CHS

CAS RN 67237-53-0

IUPAC Name: 3-ethynylthiophene
Synonyms: 3-ethynylthiophene
Molecular Weight (g/mol): 108.16
Molecular Formula: C6H4S
InChi Key: MJHLPKWONJUCFK-UHFFFAOYSA-N
SMILES: C#CC1=CSC=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (6)
  • (3)
  • (3)