missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylthiophene 97.0+%, TCI America™

Catalog No. E08925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08925G 5 g
E08921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08925G Supplier TCI America Supplier No. E08925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67237-53-0
Molecular Formula C6H4S
Molecular Weight (g/mol) 108.158
MDL Number MFCD04039973
InChI Key MJHLPKWONJUCFK-UHFFFAOYSA-N
Synonym 3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b
PubChem CID 3548422
IUPAC Name 3-ethynylthiophene
SMILES C#CC1=CSC=C1

Specifications

Color Yellow
Boiling Point 50°C
Quantity 5 g
UN Number 1993
Formula Weight 108.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethynylthiophene
TSCA No
Recommended Storage Refrigerator