missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethylthiophene 98.0+%, TCI America™
$357.90 - $1243.32
Chemical Identifiers
| CAS | 1795-01-3 |
|---|---|
| Molecular Formula | C6H8S |
| Molecular Weight (g/mol) | 112.19 |
| MDL Number | MFCD00070514 |
| InChI Key | SLDBAXYJAIRQMX-UHFFFAOYSA-N |
| PubChem CID | 74530 |
| IUPAC Name | 3-ethylthiophene |
| SMILES | CCC1=CSC=C1 |