accessibility menu, dialog, popup

UserName

H3CS

CAS RN 1795-01-3

IUPAC Name: 3-ethylthiophene
Synonyms: ethylthiophene
Molecular Weight (g/mol): 112.19
Molecular Formula: C6H8S
InChi Key: SLDBAXYJAIRQMX-UHFFFAOYSA-N
SMILES: CCC1=CSC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results