missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylthiophene 98.0+%, TCI America™

Catalog No. E03785ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03785ML 5 mL
E037825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03785ML Supplier TCI America Supplier No. E03785ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1795-01-3
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
MDL Number MFCD00070514
InChI Key SLDBAXYJAIRQMX-UHFFFAOYSA-N
PubChem CID 74530
IUPAC Name 3-ethylthiophene
SMILES CCC1=CSC=C1

Specifications

Color Yellow
Boiling Point 141°C
Quantity 5 mL
UN Number 1993
Formula Weight 112.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethylthiophene