missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylrhodanine 98.0+%, TCI America™

Catalog No. E01745G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E01745G 5 g
E017425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E01745G Supplier TCI America Supplier No. E01745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7648-01-3
Molecular Formula C5H7NOS2
Molecular Weight (g/mol) 161.24
MDL Number MFCD00051339
InChI Key UPCYEFFISUGBRW-UHFFFAOYSA-N
Synonym 3-ethylrhodanine, n-ethylrhodanine, 4-thiazolidinone, 3-ethyl-2-thioxo, rhodanine, 3-ethyl, 3-ethyl-2-thioxothiazolidin-4-one, 3-ethylrodanin, 3-ethyl-2-thioxo-4-thiazolidinone, 3-ethylrhodamine, 3-ethyl-2-thioxo-1,3-thiazolidin-4-one, unii-42klq911ht
PubChem CID 82098
IUPAC Name 3-ethyl-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES CCN1C(=O)CSC1=S

Specifications

Melting Point 37°C
Color Yellow
Boiling Point 128°C
Quantity 5 g
Formula Weight 161.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Ethylrhodanine
EINECSNumber (5)-0222
RTECSNumber VI8092000
TSCA Yes
Recommended Storage Refrigerator