accessibility menu, dialog, popup

UserName

CAS RN 7648-01-3

IUPAC Name: 3-ethyl-2-sulfanylidene-1,3-thiazolidin-4-one
Synonyms: 3-ethyl-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol): 161.24
Molecular Formula: C5H7NOS2
InChi Key: UPCYEFFISUGBRW-UHFFFAOYSA-N
SMILES: CCN1C(=O)CSC1=S
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)