missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylrhodanine, 98%

Catalog No. AAB2304706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2304706 5 g
AAB2304714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2304706 Supplier Thermo Scientific Chemicals Supplier No. B2304706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7648-01-3 | C7H3Br2F3 | 303.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7648-01-3

Specifications

Melting Point 36°C to 39°C
Density 1.303
Boiling Point 127°C to 129°C (4 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C7H3Br2F3
MDL Number MFCD00013554
Quantity 5 g
Synonym 1,4-dibromo-2-trifluoromethyl benzene, 2,5-dibromobenzotrifluoride, benzene, 1,4-dibromo-2-trifluoromethyl, 2,5-dibromo trifluoromethyl benzene, 1,4-dibromo-2-trifluoromethyl-benzene, toluene, 2,5-dibromo-alpha,alpha,alpha-trifluoro, pubchem9537, 2,5-dibromobenzotrifloride, 2,5-dibromo-, ,-trifluorotoluene
InChI Key VWKFJAOCLPPQGR-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CC(Br)=CC=C1Br
IUPAC Name 1,4-dibromo-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 303.90
PubChem CID 82108
Formula Weight 161.25
Percent Purity 98%
Chemical Name or Material 3-Ethylrhodanine
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 231-603-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only