missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium Bromide 98.0+%, TCI America™

Supplier:  TCI America E051125G

 View more versions of this product

Catalog No. E051125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium Bromide
84°C
MFCD00040549
3-ethyl-5-2-hydroxyethyl-4-methylthiazolium bromide, 3-ethyl-5-2-hydroxyethyl-4-methylthiazoliumbromide, 3-ethyl-5-2-hydroxyethyl-4-methylthiazol-3-ium bromide, thiazolium, 3-ethyl-5-2-hydroxyethyl-4-methyl-, bromide, 2-3-ethyl-4-methyl-thiazol-3-ium-5-yl ethanol bromide, 3-ethyl-5-2-hydroxyethyl-4-methyl-1,3-thiazol-3-ium bromide, pubchem15236, acmc-1area, ksc272q3h, 3ethyl-5-2-hydroxyethyl-4-methylthiazolium bromide
CC[N+]1=CSC(=C1C)CCO.[Br-]
252.17
252.17
Crystalline Powder
54016-70-5
C8H14BrNOS
25 g
BDQRQMLWZJQQKS-UHFFFAOYSA-M
2-(3-ethyl-4-methyl-1,3-thiazol-3-ium-5-yl)ethanol;bromide
9964923
≥98.0% (HPLC,T)

Chemical Identifiers

54016-70-5
252.17
BDQRQMLWZJQQKS-UHFFFAOYSA-M
9964923
CC[N+]1=CSC(=C1C)CCO.[Br-]
C8H14BrNOS
MFCD00040549
3-ethyl-5-2-hydroxyethyl-4-methylthiazolium bromide, 3-ethyl-5-2-hydroxyethyl-4-methylthiazoliumbromide, 3-ethyl-5-2-hydroxyethyl-4-methylthiazol-3-ium bromide, thiazolium, 3-ethyl-5-2-hydroxyethyl-4-methyl-, bromide, 2-3-ethyl-4-methyl-thiazol-3-ium-5-yl ethanol bromide, 3-ethyl-5-2-hydroxyethyl-4-methyl-1,3-thiazol-3-ium bromide, pubchem15236, acmc-1area, ksc272q3h, 3ethyl-5-2-hydroxyethyl-4-methylthiazolium bromide
2-(3-ethyl-4-methyl-1,3-thiazol-3-ium-5-yl)ethanol;bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions