missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium Bromide 98.0+%, TCI America™

Catalog No. p-7129040
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05115G 5 g
E051125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05115G Supplier TCI America Supplier No. E05115G
Only null left

Chemical Identifiers

CAS 54016-70-5
Molecular Formula C8H14BrNOS
Molecular Weight (g/mol) 252.17
MDL Number MFCD00040549
InChI Key BDQRQMLWZJQQKS-UHFFFAOYSA-M
Synonym 3-ethyl-5-2-hydroxyethyl-4-methylthiazolium bromide, 3-ethyl-5-2-hydroxyethyl-4-methylthiazoliumbromide, 3-ethyl-5-2-hydroxyethyl-4-methylthiazol-3-ium bromide, thiazolium, 3-ethyl-5-2-hydroxyethyl-4-methyl-, bromide, 2-3-ethyl-4-methyl-thiazol-3-ium-5-yl ethanol bromide, 3-ethyl-5-2-hydroxyethyl-4-methyl-1,3-thiazol-3-ium bromide, pubchem15236, acmc-1area, ksc272q3h, 3ethyl-5-2-hydroxyethyl-4-methylthiazolium bromide
PubChem CID 9964923
IUPAC Name 2-(3-ethyl-4-methyl-1,3-thiazol-3-ium-5-yl)ethanol;bromide
SMILES CC[N+]1=CSC(=C1C)CCO.[Br-]

Specifications

Melting Point 84°C
Quantity 5 g
Formula Weight 252.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium Bromide