missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-4-methyl-3-pyrrolin-2-one 98.0+%, TCI America™

Supplier:  TCI America E096425G

 View more versions of this product

Catalog No. E096425G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Ethyl-4-methyl-3-pyrrolin-2-one
104°C
C7H11NO
25 g
YCTNTSVMJWIYTQ-UHFFFAOYSA-N
3-ethyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one
854146
≥98.0% (HPLC,N)
766-36-9
White-Yellow
MFCD00173861
3-ethyl-4-methyl-3-pyrrolin-2-one, 3-ethyl-4-methyl-3-pyrroline-2-one, 3-ethyl-4-methyl-2,5-dihydro-1h-pyrrol-2-one, 3-ethyl-4-methyl-1h-pyrrol-2 5h-one, 1,5-dihydro-3-ethyl-4-methyl-2h-pyrrol-2-one, 2h-pyrrol-2-one, 3-ethyl-1,5-dihydro-4-methyl, 3-ethyl-4-methyl-2-oxo-3-pyrroline, 3-ethyl-4-methyl-1,5-dihydropyrrol-2-one, 3-ethyl-1,5-dihydro-4-methyl-2h-pyrrol-2-one, pubchem12392
CCC1=C(C)CNC1=O
125.17
125.17
Crystalline Powder

Chemical Identifiers

766-36-9
125.17
YCTNTSVMJWIYTQ-UHFFFAOYSA-N
854146
CCC1=C(C)CNC1=O
C7H11NO
MFCD00173861
3-ethyl-4-methyl-3-pyrrolin-2-one, 3-ethyl-4-methyl-3-pyrroline-2-one, 3-ethyl-4-methyl-2,5-dihydro-1h-pyrrol-2-one, 3-ethyl-4-methyl-1h-pyrrol-2 5h-one, 1,5-dihydro-3-ethyl-4-methyl-2h-pyrrol-2-one, 2h-pyrrol-2-one, 3-ethyl-1,5-dihydro-4-methyl, 3-ethyl-4-methyl-2-oxo-3-pyrroline, 3-ethyl-4-methyl-1,5-dihydropyrrol-2-one, 3-ethyl-1,5-dihydro-4-methyl-2h-pyrrol-2-one, pubchem12392
3-ethyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions