missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-4-methyl-3-pyrrolin-2-one 98.0+%, TCI America™

Catalog No. p-7129249
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09645G 5 g
E096425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09645G Supplier TCI America Supplier No. E09645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 766-36-9
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.17
MDL Number MFCD00173861
InChI Key YCTNTSVMJWIYTQ-UHFFFAOYSA-N
Synonym 3-ethyl-4-methyl-3-pyrrolin-2-one, 3-ethyl-4-methyl-3-pyrroline-2-one, 3-ethyl-4-methyl-2,5-dihydro-1h-pyrrol-2-one, 3-ethyl-4-methyl-1h-pyrrol-2 5h-one, 1,5-dihydro-3-ethyl-4-methyl-2h-pyrrol-2-one, 2h-pyrrol-2-one, 3-ethyl-1,5-dihydro-4-methyl, 3-ethyl-4-methyl-2-oxo-3-pyrroline, 3-ethyl-4-methyl-1,5-dihydropyrrol-2-one, 3-ethyl-1,5-dihydro-4-methyl-2h-pyrrol-2-one, pubchem12392
PubChem CID 854146
IUPAC Name 3-ethyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one
SMILES CCC1=C(C)CNC1=O

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 5 g
Formula Weight 125.17
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethyl-4-methyl-3-pyrrolin-2-one