missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-pentanol 99.0+%, TCI America™

Supplier:  TCI America E0157100ML

 View more versions of this product

Catalog No. E0157100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Ethyl-3-pentanol
Colorless
C7H16O
100 mL
3-ethyl-3-pentanol, triethylcarbinol, 3-pentanol, 3-ethyl, triethylmethanol, triethylmethyl alcohol, tert-heptanol, 3-aethyl-pentanol-3, 3-aethyl-pentanol-3 german, 3-ethyl-3-hydroxypentane, ethyl-3 pentanol-3
CCC(O)(CC)CC
116.20
116.20
Liquid
597-49-9
143°C
MFCD00004484
1987
XKIRHOWVQWCYBT-UHFFFAOYSA-N
3-ethylpentan-3-ol
11702
≥99.0% (GC)

Chemical Identifiers

597-49-9
116.20
XKIRHOWVQWCYBT-UHFFFAOYSA-N
11702
CCC(O)(CC)CC
C7H16O
MFCD00004484
3-ethyl-3-pentanol, triethylcarbinol, 3-pentanol, 3-ethyl, triethylmethanol, triethylmethyl alcohol, tert-heptanol, 3-aethyl-pentanol-3, 3-aethyl-pentanol-3 german, 3-ethyl-3-hydroxypentane, ethyl-3 pentanol-3
3-ethylpentan-3-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

RTECSNumber : SA6780000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions