missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-pentanol 99.0+%, TCI America™

Catalog No. p-7128867
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E015725ML 25 mL
E0157100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E015725ML Supplier TCI America Supplier No. E015725ML

Chemical Identifiers

CAS 597-49-9
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00004484
InChI Key XKIRHOWVQWCYBT-UHFFFAOYSA-N
Synonym 3-ethyl-3-pentanol, triethylcarbinol, 3-pentanol, 3-ethyl, triethylmethanol, triethylmethyl alcohol, tert-heptanol, 3-aethyl-pentanol-3, 3-aethyl-pentanol-3 german, 3-ethyl-3-hydroxypentane, ethyl-3 pentanol-3
PubChem CID 11702
IUPAC Name 3-ethylpentan-3-ol
SMILES CCC(O)(CC)CC

Specifications

Color Colorless
Boiling Point 143°C
Quantity 25 mL
UN Number 1987
Formula Weight 116.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethyl-3-pentanol
EINECSNumber (2)-0217
RTECSNumber SA6780000
TSCA Yes