missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-methylglutarimide 98.0+%, TCI America™

Catalog No. E028410G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E028410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E028410G Supplier TCI America Supplier No. E028410G
Only null left

Chemical Identifiers

CAS 64-65-3
Molecular Formula C8H13NO2
Molecular Weight (g/mol) 155.197
MDL Number MFCD00006673
InChI Key ORRZGUBHBVWWOP-UHFFFAOYSA-N
PubChem CID 2310
IUPAC Name 4-ethyl-4-methylpiperidine-2,6-dione
SMILES CCC1(CC(=O)NC(=O)C1)C

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 10 g
UN Number 2811
Formula Weight 155.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethyl-3-methylglutarimide
RTECSNumber MA4550000
TSCA Yes