missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-methylpentane 99.0+%, TCI America™

Catalog No. E02595ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02595ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E02595ML Supplier TCI America Supplier No. E02595ML
Only null left

Chemical Identifiers

CAS 609-26-7
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00048665
InChI Key DUPUVYJQZSLSJB-UHFFFAOYSA-N
PubChem CID 11863
IUPAC Name 3-ethyl-2-methylpentane
SMILES CCC(CC)C(C)C

Specifications

Color Colorless
Boiling Point 116°C
Quantity 5 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethyl-2-methylpentane
EINECSNumber (2)-0008
TSCA No