accessibility menu, dialog, popup

UserName

H3CCH3CH3H3C

CAS RN 609-26-7

IUPAC Name: 3-ethyl-2-methylpentane
Synonyms: 2-methyl-3-ethylpentane
Molecular Weight (g/mol): 114.23
Molecular Formula: C8H18
InChi Key: DUPUVYJQZSLSJB-UHFFFAOYSA-N
SMILES: CCC(CC)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results