missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethyl-2-methylbenzoxazolium Iodide 98.0+%, TCI America™
Supplier: TCI America E047725G
Specifications
| 3-Ethyl-2-methylbenzoxazolium Iodide | |
| 197°C | |
| MFCD00044141 | |
| 3-ethyl-2-methylbenzoxazolium iodide, 3-ethyl-2-methylbenzo d oxazol-3-ium iodide, 2-methyl-3-ethylbenzoxazolium iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide 1:1, 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide, 3-ethyl-2-methylbenzoxazoliumiodide, acmc-209l0h, ethyl-2-methyl-benzoxazolium iodide, 3-ethyl-2-methylbenzoxazolium iodide, technical grade | |
| [I-].CC[N+]1=C(C)OC2=CC=CC=C12 | |
| 289.12 | |
| 289.12 | |
| Crystalline Powder |
| 5260-37-7 | |
| C10H12INO | |
| 25 g | |
| UZWYLWSUBQEUMC-UHFFFAOYSA-M | |
| 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide | |
| 21331 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 5260-37-7 | |
| 289.12 | |
| UZWYLWSUBQEUMC-UHFFFAOYSA-M | |
| 21331 | |
| [I-].CC[N+]1=C(C)OC2=CC=CC=C12 |
| C10H12INO | |
| MFCD00044141 | |
| 3-ethyl-2-methylbenzoxazolium iodide, 3-ethyl-2-methylbenzo d oxazol-3-ium iodide, 2-methyl-3-ethylbenzoxazolium iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide 1:1, 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide, 3-ethyl-2-methylbenzoxazoliumiodide, acmc-209l0h, ethyl-2-methyl-benzoxazolium iodide, 3-ethyl-2-methylbenzoxazolium iodide, technical grade | |
| 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide |
Safety and Handling
EINECSNumber : (5)-0159
RTECSNumber : DM5285000
TSCA : Yes