accessibility menu, dialog, popup

UserName

CAS RN 5260-37-7

IUPAC Name: 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide
Synonyms: 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide
Molecular Weight (g/mol): 289.12
Molecular Formula: C10H12INO
InChi Key: UZWYLWSUBQEUMC-UHFFFAOYSA-M
SMILES: [I-].CC[N+]1=C(C)OC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results