missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethyl-2-methyl-3-pentanol 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 59°C |
| Quantity | 5 g |
| UN Number | 1987 |
| Formula Weight | 130.23 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Ethyl-2-methyl-3-pentanol |
Safety and Handling
| EINECSNumber | (2)-0217 |
| TSCA | No |