accessibility menu, dialog, popup

UserName

CH3OHCH3CH3H3C

CAS RN 597-05-7

IUPAC Name: 3-ethyl-2-methylpentan-3-ol
Synonyms: 3-pentanol, 3-ethyl-2-methyl-
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: DMHIJUVUPKCGLJ-UHFFFAOYSA-N
SMILES: CCC(O)(CC)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results