missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-methyl-1-pentene, 99%, Thermo Scientific Chemicals

Catalog No. AAB2062406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2062406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2062406 Supplier Thermo Scientific Chemicals Supplier No. B2062406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19780-66-6 | Molecular Formula: C8H16 | Molecular Weight: 112.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19780-66-6
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00048664
InChI Key HPHHYSWOBXEIRG-UHFFFAOYSA-N
Synonym 3-ethyl-2-methyl-1-pentene, 1-pentene, 3-ethyl-2-methyl, 2-methyl-3-ethyl-1-pentene, acmc-1bqij, 1-pentene,3-ethyl-2-methyl
PubChem CID 140591
IUPAC Name 3-ethyl-2-methylpent-1-ene
SMILES CCC(CC)C(=C)C

Specifications

Density 0.72
Refractive Index 1.4124
Quantity 5 g
UN Number UN3295
Formula Weight 112.22
Percent Purity 99%
Chemical Name or Material 3-Ethyl-2-methyl-1-pentene
Hazard Category H225
Hazard Statement GHS H Statement
H224
Extremely flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only