CAS RN 19780-66-6
IUPAC Name:
3-ethyl-2-methylpent-1-ene
Synonyms:
1-pentene, 3-ethyl-2-methyl-
Molecular Weight (g/mol):
112.22
Molecular Formula:
C8H16
InChi Key:
HPHHYSWOBXEIRG-UHFFFAOYSA-N
SMILES:
CCC(CC)C(C)=C
1
–
1
of
1
results
1
Promotions Available
3-Ethyl-2-methyl-1-pentene, 99%, Thermo Scientific Chemicals
3-Ethyl-2-methyl-1-pentene, 99%, Thermo Scientific Chemicals
CAS: 19780-66-6 Molecular Formula: C8H16 Molecular Weight (g/mol): 112.216 MDL Number: MFCD00048664 InChI Key: HPHHYSWOBXEIRG-UHFFFAOYSA-N Synonym: 3-ethyl-2-methyl-1-pentene,1-pentene, 3-ethyl-2-methyl,2-methyl-3-ethyl-1-pentene,acmc-1bqij,1-pentene,3-ethyl-2-methyl PubChem CID: 140591 IUPAC Name: 3-ethyl-2-methylpent-1-ene SMILES: CCC(CC)C(=C)C