missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-hydroxy-2-cyclopenten-1-one 97.0+%, TCI America™

Catalog No. E07925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07925G 5 g
E079225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07925G Supplier TCI America Supplier No. E07925G

Chemical Identifiers

CAS 21835-01-8
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00012391
InChI Key JHWFWLUAUPZUCP-UHFFFAOYSA-N
PubChem CID 62752
IUPAC Name 3-ethyl-2-hydroxycyclopent-2-en-1-one
SMILES CCC1=C(C(=O)CC1)O

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 75°C
Quantity 5 g
Formula Weight 126.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethyl-2-hydroxy-2-cyclopenten-1-one
TSCA Yes
Recommended Storage Freezer