accessibility menu, dialog, popup

UserName

H3COHO

CAS RN 21835-01-8

IUPAC Name: 3-ethyl-2-hydroxycyclopent-2-en-1-one
Synonyms: 3-ethyl-2-hydroxycyclopent-2-en-1-one
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: JHWFWLUAUPZUCP-UHFFFAOYSA-N
SMILES: CCC1=C(O)C(=O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results