Learn More
3-(Ethoxycarbonyl)phenyl isocyanate, 97%
CAS: 67531-68-4 | C10H9NO3 | 191.19 g/mol
Supplier: Thermo Scientific Chemicals L0949506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-(Ethoxycarbonyl)phenyl isocyanate | |
| 1.154 | |
| >110°C (230°F) | |
| 1.531 | |
| 5 g | |
| 7636626 | |
| 3-ethoxycarbonyl phenyl isocyanate, 3-carbethoxyphenyl isocyanate, 3-ethoxycarbonylphenyl isocyanate, 3-ethoxycarbonyl phenylisocyanate, 3-isocyanatobenzoic acid ethyl ester, 3-carboethoxyphenyl isocyanate, #, ethyl 3-isocyanatobenzoate, 3-ethoxycarbonyl phenylisothiocyanate, 3-isocyanato-benzoic acid ethyl ester | |
| CCOC(=O)C1=CC(=CC=C1)N=C=O | |
| 191.19 | |
| 191.19 |
| 67531-68-4 | |
| 213°C | |
| C10H9NO3 | |
| MFCD00013865 | |
| UN2206 | |
| Moisture sensitive | |
| UPXGGIRFDZENRD-UHFFFAOYSA-N | |
| ethyl 3-isocyanatobenzoate | |
| 589806 | |
| 97% |
Chemical Identifiers
| 67531-68-4 | |
| 191.19 | |
| UPXGGIRFDZENRD-UHFFFAOYSA-N | |
| 589806 | |
| CCOC(=O)C1=CC(=CC=C1)N=C=O |
| C10H9NO3 | |
| MFCD00013865 | |
| 3-ethoxycarbonyl phenyl isocyanate, 3-carbethoxyphenyl isocyanate, 3-ethoxycarbonylphenyl isocyanate, 3-ethoxycarbonyl phenylisocyanate, 3-isocyanatobenzoic acid ethyl ester, 3-carboethoxyphenyl isocyanate, #, ethyl 3-isocyanatobenzoate, 3-ethoxycarbonyl phenylisothiocyanate, 3-isocyanato-benzoic acid ethyl ester | |
| ethyl 3-isocyanatobenzoate |
Safety and Handling
GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H331-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only