missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxybenzaldehyde, 97%

CAS: 22924-15-8 | C9H10O2 | 150.177 g/mol

$144.92 - $436.03

Chemical Identifiers

CAS 22924-15-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00016606
InChI Key QZMGMXBYJZVAJN-UHFFFAOYSA-N
Synonym m-ethoxybenzaldehyde, benzaldehyde, 3-ethoxy, 3-ethoxy-benzaldehyde, benzaldehyde, m-ethoxy, 3-ethoxy benzaldehyde, 3-ethoxybenzaldehdye, pubchem13132, acmc-209fzo, intermediates-zcf02615, 3-ethoxybenzaldehyde
PubChem CID 89908
IUPAC Name 3-ethoxybenzaldehyde
SMILES CCOC1=CC=CC(=C1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1894088
View Documents
Thermo Scientific Chemicals
A1894088
2.5 g
Each for $144.92
Only null left
 
AAA1894009
View Documents
Thermo Scientific Chemicals
A1894009
10 g
Each for $436.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22924-15-8
150.177
QZMGMXBYJZVAJN-UHFFFAOYSA-N
89908
CCOC1=CC=CC(=C1)C=O
C9H10O2
MFCD00016606
m-ethoxybenzaldehyde, benzaldehyde, 3-ethoxy, 3-ethoxy-benzaldehyde, benzaldehyde, m-ethoxy, 3-ethoxy benzaldehyde, 3-ethoxybenzaldehdye, pubchem13132, acmc-209fzo, intermediates-zcf02615, 3-ethoxybenzaldehyde
3-ethoxybenzaldehyde

Specifications

22924-15-8
132°C to 134°C (15 mmHg)
MFCD00016606
Air Sensitive
QZMGMXBYJZVAJN-UHFFFAOYSA-N
3-ethoxybenzaldehyde
89908
97%
1.07
C9H10O2
2.5 g
m-ethoxybenzaldehyde, benzaldehyde, 3-ethoxy, 3-ethoxy-benzaldehyde, benzaldehyde, m-ethoxy, 3-ethoxy benzaldehyde, 3-ethoxybenzaldehdye, pubchem13132, acmc-209fzo, intermediates-zcf02615, 3-ethoxybenzaldehyde
CCOC1=CC=CC(=C1)C=O
150.177
150.18
3-Ethoxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 245-333-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only