missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethoxyacrylonitrile (cis- and trans- mixture) 98.0+%, TCI America™
$119.30 - $892.89
Chemical Identifiers
| CAS | 61310-53-0 |
|---|---|
| Molecular Formula | C5H7NO |
| Molecular Weight (g/mol) | 97.12 |
| MDL Number | MFCD00010193 |
| InChI Key | HUPVIAINOSTNBJ-HYXAFXHYSA-N |
| Synonym | 2z-3-ethoxyprop-2-enenitrile, z-3-ethoxyprop-2-enenitrile, 2-propenenitrile, 3-ethoxy, z-3-ethoxy-2-propenenitrile |
| PubChem CID | 5324714 |
| IUPAC Name | (2Z)-3-ethoxyprop-2-enenitrile |
| SMILES | CCO\C=C/C#N |
Chemical Identifiers
| 61310-53-0 | |
| 97.12 | |
| HUPVIAINOSTNBJ-HYXAFXHYSA-N | |
| 5324714 | |
| CCO\C=C/C#N |
| C5H7NO | |
| MFCD00010193 | |
| 2z-3-ethoxyprop-2-enenitrile, z-3-ethoxyprop-2-enenitrile, 2-propenenitrile, 3-ethoxy, z-3-ethoxy-2-propenenitrile | |
| (2Z)-3-ethoxyprop-2-enenitrile |
Specifications
| 61310-53-0 | |
| 71°C | |
| MFCD00010193 | |
| 2z-3-ethoxyprop-2-enenitrile, z-3-ethoxyprop-2-enenitrile, 2-propenenitrile, 3-ethoxy, z-3-ethoxy-2-propenenitrile | |
| CCO\C=C/C#N | |
| 97.12 | |
| 97.12 | |
| Liquid |
| Yellow | |
| C5H7NO | |
| 25 mL | |
| HUPVIAINOSTNBJ-HYXAFXHYSA-N | |
| (2Z)-3-ethoxyprop-2-enenitrile | |
| 5324714 | |
| ≥98.0% (GC) | |
| 3-Ethoxyacrylonitrile (cis- and trans- mixture) |
Safety and Handling
RTECSNumber : AT6475000
TSCA : No